Assay Details
| Gene Symbol: Atp5mc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001305067.1 | NP_001291996.1 | 2 - 3 | 62 | 86 | |
| GenBank mRNA | KM486687.1 | - | 2 - 3 | 62 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001305067.1 transcripts.
Set Membership: |
|

Atp5mc1
Genomic Map