Assay Details
| Gene Symbol: Hif1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310267.1 | NP_001297196.1 | 7 - 8 | 1241 | 71 | |
| XM_004837433.3 | XP_004837490.1 | 7 - 8 | 986 | 71 | ||
| GenBank mRNA | KM486721.1 | - | 7 - 8 | 1241 | 71 | |
More Information
Set Membership: |
|

Hif1a
Genomic Map