Assay Details
| Gene Symbol: Rpl37a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267853.2 | NP_001254782.1 | 1 - 2 | 50 | 71 | |
| GenBank mRNA | HQ326676.2 | - | 1 - 2 | 50 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001267853.2 transcripts.
Set Membership: |
|

Rpl37a
Genomic Map