Assay Details
| Gene Symbol: Tomt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001282110.1 | NP_001269039.1 | 1 - 2 | 257 | 66 | |
| XM_021243175.1 | XP_021098834.1 | 3 - 4 | 1134 | 66 | ||
| XM_021243176.1 | XP_021098835.1 | 2 - 3 | 432 | 66 | ||
More Information
Set Membership: |
|

Tomt
Genomic Map