Assay Details
| Gene Symbol: Clk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310289.1 | NP_001297218.1 | 9 - 10 | 1243 | 67 | |
| XM_004871274.3 | XP_004871331.1 | 9 - 10 | 1245 | 67 | ||
| XM_021249251.1 | XP_021104910.1 | 9 - 10 | 1244 | 67 | ||
| XM_021249252.1 | XP_021104911.1 | 9 - 10 | 1242 | 67 | ||
| GenBank mRNA | KM486693.1 | - | 9 - 10 | 1243 | 67 | |
More Information
Set Membership: |
|

Clk2
Genomic Map