Assay Details
| Gene Symbol: Polr2d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001279871.1 | NP_001266800.1 | 2 - 3 | 273 | 68 | |
| XM_013075570.2 | XP_012931024.1 | 2 - 3 | 354 | 68 | ||
| GenBank mRNA | HQ326674.2 | - | 2 - 3 | 274 | 68 | |
More Information
Set Membership: |
|

Polr2d
Genomic Map