Assay Details
| Gene Symbol: Aco2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001308662.1 | NP_001295591.1 | 10 - 11 | 1311 | 52 | |
| GenBank mRNA | KM486681.1 | - | 10 - 11 | 1312 | 52 | |
More Information
Set Membership: |
|

Aco2
Genomic Map