Assay Details
| Gene Symbol: Ran | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001310259.1 | NP_001297188.1 | 3 - 4 | 332 | 66 | |
| GenBank mRNA | KM486760.1 | - | 3 - 4 | 332 | 66 | |
More Information
Set Membership: |
|

Ran
Genomic Map