Assay Details
Gene Symbol: Ugt8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870034.2 | XP_004870091.1 | 4 - 5 | 1276 | 71 | |
XM_004870035.2 | XP_004870092.1 | 4 - 5 | 1301 | 71 | ||
XM_021248465.1 | XP_021104124.1 | 5 - 6 | 1346 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
