Assay Details
| Gene Symbol: Dnase2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872396.3 | XP_004872453.1 | 5 - 6 | 864 | 67 | |
| XM_004872398.2 | XP_004872455.1 | 5 - 6 | 821 | 67 | ||
More Information
Set Membership: |
|

Dnase2
Genomic Map