Assay Details
| Gene Symbol: LOC101708169 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864145.3 | XP_004864202.1 | 1 - 2 | 229 | 59 | |
| XM_021243878.1 | XP_021099537.1 | 1 - 2 | 235 | 59 | ||
More Information
Set Membership: |
|

LOC101708169
Genomic Map