Assay Details
| Gene Symbol: Elovl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862898.3 | XP_004862955.1 | 5 - 6 | 542 | 97 | |
| XM_004862900.2 | XP_004862957.1 | 3 - 4 | 276 | 97 | ||
| XM_021242942.1 | XP_021098601.1 | 6 - 7 | 688 | 97 | ||
| XM_021242943.1 | XP_021098602.1 | 4 - 5 | 467 | 97 | ||
More Information
Set Membership: |
|

Elovl7
Genomic Map