Assay Details
| Gene Symbol: Serpinf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004857147.3 | XP_004857204.1 | 4 - 5 | 652 | 62 | |
| XM_021238181.1 | XP_021093840.1 | 3 - 4 | 441 | 62 | ||
More Information
Set Membership: |
|

Serpinf1
Genomic Map