Assay Details
| Gene Symbol: Prdx4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872215.2 | XP_004872272.1 | 4 - 5 | 808 | 90 | |
| XM_004872216.2 | XP_004872273.1 | 4 - 5 | 662 | 90 | ||
| XM_013068316.2 | XP_012923770.1 | 4 - 5 | 809 | 90 | ||
| XM_021250196.1 | XP_021105855.1 | 4 - 5 | 809 | 90 | ||
More Information
Set Membership: |
|

Prdx4
Genomic Map