Assay Details
| Gene Symbol: Fcn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850708.3 | XP_004850765.1 | 7 - 8 | 725 | 66 | |
| XM_004850709.3 | XP_004850766.1 | 6 - 7 | 692 | 66 | ||
| XM_021263143.1 | XP_021118802.1 | 7 - 8 | 786 | 66 | ||
| XM_021263144.1 | XP_021118803.1 | 7 - 8 | 783 | 66 | ||
| XM_021263145.1 | XP_021118804.1 | 6 - 7 | 754 | 66 | ||
More Information
Set Membership: |
|

Fcn3
Genomic Map