Assay Details
| Gene Symbol: Dnajc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859365.3 | XP_004859422.2 | 5 - 6 | 543 | 58 | |
| XM_004859366.2 | XP_004859423.1 | 5 - 6 | 795 | 58 | ||
More Information
Set Membership: |
|

Dnajc7
Genomic Map