Assay Details
| Gene Symbol: LOC101717359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836997.3 | XP_004837054.1 | 1 - 2 | 123 | 92 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004836997.3 transcripts.
Set Membership: |
|

LOC101717359
Genomic Map