Assay Details
| Gene Symbol: Naa50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835702.3 | XP_004835759.1 | 2 - 3 | 427 | 63 | |
| XM_021263581.1 | XP_021119240.1 | 2 - 3 | 425 | 63 | ||
More Information
Set Membership: |
|

Naa50
Genomic Map