Assay Details
| Gene Symbol: Ebpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844153.2 | XP_004844210.1 | 3 - 4 | 464 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ebpl| Gene Symbol: Ebpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844153.2 | XP_004844210.1 | 3 - 4 | 464 | 101 | |
Set Membership: |
|