Assay Details
| Gene Symbol: Rnf7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834362.3 | XP_004834419.1 | 2 - 3 | 330 | 96 | |
| XM_004834364.3 | XP_004834421.1 | 2 - 3 | 468 | 96 | ||
More Information
Set Membership: |
|

Rnf7
Genomic Map