Assay Details
| Gene Symbol: Hsd17b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004861272.3 | XP_004861329.1 | 1 - 2 | 205 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004861272.3 transcripts.
Set Membership: |
|

Hsd17b6
Genomic Map