Assay Details
| Gene Symbol: Srprb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834256.3 | XP_004834313.1 | 2 - 3 | 303 | 64 | |
| XM_021244916.1 | XP_021100575.1 | 2 - 3 | 290 | 64 | ||
More Information
Set Membership: |
|

Srprb
Genomic Map