Assay Details
| Gene Symbol: Psma5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853629.2 | XP_004853686.1 | 6 - 7 | 599 | 78 | |
| XM_021265611.1 | XP_021121270.1 | 7 - 8 | 658 | 78 | ||
| XM_021265612.1 | XP_021121271.1 | 7 - 8 | 625 | 78 | ||
| XM_021265613.1 | XP_021121272.1 | 5 - 6 | 498 | 78 | ||
More Information
Set Membership: |
|

Psma5
Genomic Map