Assay Details
| Gene Symbol: Podxl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021237550.1 | XP_021093209.1 | 4 - 5 | 1334 | 76 | |
| XM_021237551.1 | XP_021093210.1 | 4 - 5 | 1303 | 76 | ||
| XM_021237552.1 | XP_021093211.1 | 4 - 5 | 1316 | 76 | ||
More Information
Set Membership: |
|

Podxl
Genomic Map