Assay Details
| Gene Symbol: Galk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004835432.2 | XP_004835489.1 | 8 - 9 | 1061 | 62 | |
| XM_004835433.3 | XP_004835490.1 | 8 - 9 | 998 | 62 | ||
More Information
Set Membership: |
|

Galk2
Genomic Map