Assay Details
| Gene Symbol: Tbcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846564.3 | XP_004846621.1 | 1 | 971 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tbcc| Gene Symbol: Tbcc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846564.3 | XP_004846621.1 | 1 | 971 | 96 | |
Set Membership: |
|