Assay Details
| Gene Symbol: Dnajb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841398.3 | XP_004841455.1 | 4 - 5 | 1277 | 106 | |
| XM_004841399.3 | XP_004841456.1 | 4 - 5 | 1105 | 106 | ||
More Information
Set Membership: |
|

Dnajb4
Genomic Map