Assay Details
| Gene Symbol: Pdpk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864657.3 | XP_004864714.1 | 9 - 10 | 1091 | 64 | |
| XM_021244477.1 | XP_021100136.1 | 10 - 11 | 1073 | 64 | ||
More Information
Set Membership: |
|

Pdpk1
Genomic Map