Assay Details
| Gene Symbol: Prdx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872929.3 | XP_004872986.2 | 3 - 4 | 422 | 68 | |
| XM_013068549.2 | XP_012924003.1 | 3 - 4 | 419 | 68 | ||
More Information
Set Membership: |
|

Prdx1
Genomic Map