Assay Details
| Gene Symbol: Emg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869477.2 | XP_004869534.1 | 3 - 4 | 525 | 62 | |
| XM_021248031.1 | XP_021103690.1 | 3 - 4 | 715 | 62 | ||
| XM_021248032.1 | XP_021103691.1 | 3 - 4 | 727 | 62 | ||
More Information
Set Membership: |
|

Emg1
Genomic Map