Assay Details
| Gene Symbol: Tcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869710.1 | XP_004869767.1 | 1 - 2 | 121 | 104 | |
| XM_004869711.1 | XP_004869768.1 | 1 - 2 | 121 | 104 | ||
| XM_021248336.1 | XP_021103995.1 | 1 - 2 | 121 | 104 | ||
More Information
Set Membership: |
|

Tcn1
Genomic Map