Assay Details
| Gene Symbol: Tcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869710.1 | XP_004869767.1 | 6 - 7 | 977 | 94 | |
| XM_004869711.1 | XP_004869768.1 | 5 - 6 | 821 | 94 | ||
| XM_004869712.1 | XP_004869769.1 | 5 - 6 | 797 | 94 | ||
| XM_021248336.1 | XP_021103995.1 | 6 - 7 | 974 | 94 | ||

Tcn1
Genomic Map