Assay Details
| Gene Symbol: Tcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869710.1 | XP_004869767.1 | 7 - 8 | 1163 | 70 | |
| XM_004869711.1 | XP_004869768.1 | 6 - 7 | 1007 | 70 | ||
| XM_004869712.1 | XP_004869769.1 | 6 - 7 | 983 | 70 | ||
| XM_021248336.1 | XP_021103995.1 | 7 - 8 | 1160 | 70 | ||
More Information
Set Membership: |
|

Tcn1
Genomic Map