Assay Details
| Gene Symbol: Tagap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874544.2 | XP_004874601.1 | 9 - 10 | 1242 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tagap| Gene Symbol: Tagap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004874544.2 | XP_004874601.1 | 9 - 10 | 1242 | 67 | |
Set Membership: |
|