Assay Details
| Gene Symbol: Znf207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870595.3 | XP_004870652.1 | 11 - 12 | 1478 | 69 | |
| XM_004870597.3 | XP_004870654.1 | 10 - 11 | 1430 | 69 | ||
| XM_004870599.3 | XP_004870656.1 | 10 - 11 | 1385 | 69 | ||
| XM_013067760.2 | XP_012923214.1 | 11 - 12 | 1478 | 69 | ||
| XM_013067761.2 | XP_012923215.1 | 11 - 12 | 1474 | 69 | ||
| XM_013067762.2 | XP_012923216.1 | 10 - 11 | 1426 | 69 | ||
| XM_021248877.1 | XP_021104536.1 | 10 - 11 | 1381 | 69 | ||
More Information
Set Membership: |
|

Znf207
Genomic Map