Assay Details
| Gene Symbol: Adipor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004837640.2 | XP_004837697.1 | 6 - 7 | 1112 | 56 | |
| XM_013068952.2 | XP_012924406.1 | 7 - 8 | 1191 | 56 | ||
| XM_021249805.1 | XP_021105464.1 | 7 - 8 | 1187 | 56 | ||
| XM_021249808.1 | XP_021105467.1 | 6 - 7 | 944 | 56 | ||
More Information
Set Membership: |
|

Adipor2
Genomic Map