Assay Details
| Gene Symbol: Rfc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004834814.2 | XP_004834871.1 | 6 - 7 | 807 | 80 | |
| XM_004834815.2 | XP_004834872.1 | 6 - 7 | 811 | 80 | ||
| XM_004834816.2 | XP_004834873.1 | 6 - 7 | 807 | 80 | ||
More Information
Set Membership: |
|

Rfc4
Genomic Map