Assay Details
| Gene Symbol: Fkbp1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839088.2 | XP_004839145.1 | 1 - 2 | 199 | 65 | |
| XM_021253548.1 | XP_021109207.1 | 1 - 2 | 209 | 65 | ||
More Information
Set Membership: |
|

Fkbp1b
Genomic Map