Assay Details
| Gene Symbol: Mbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865938.2 | XP_004865995.1 | 5 - 6 | 730 | 66 | |
| XM_004865939.3 | XP_004865996.1 | 5 - 6 | 659 | 66 | ||
More Information
Set Membership: |
|

Mbd3
Genomic Map