Assay Details
| Gene Symbol: Cpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262674.1 | XP_021118333.1 | 6 - 7 | 1725 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpq| Gene Symbol: Cpq | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262674.1 | XP_021118333.1 | 6 - 7 | 1725 | 65 | |
Set Membership: |
|