Assay Details
| Gene Symbol: Ormdl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859477.3 | XP_004859534.1 | 2 - 3 | 320 | 70 | |
| XM_021240079.1 | XP_021095738.1 | 2 - 3 | 305 | 70 | ||
More Information
Set Membership: |
|

Ormdl3
Genomic Map