Assay Details
| Gene Symbol: Tomm20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854628.3 | XP_004854685.1 | 2 - 3 | 411 | 101 | |
| XM_004854629.3 | XP_004854686.1 | 2 - 3 | 390 | 101 | ||
| XM_021266393.1 | XP_021122052.1 | 2 - 3 | 410 | 101 | ||
More Information
Set Membership: |
|

Tomm20
Genomic Map