Assay Details
| Gene Symbol: Atp6v1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856373.3 | XP_004856430.1 | 1 - 2 | 206 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp6v1f| Gene Symbol: Atp6v1f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004856373.3 | XP_004856430.1 | 1 - 2 | 206 | 100 | |
Set Membership: |
|