Assay Details
| Gene Symbol: Larp6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004855551.2 | XP_004855608.1 | 2 - 3 | 511 | 72 | |
| XM_004855552.2 | XP_004855609.1 | 2 - 3 | 505 | 72 | ||
| XM_021236990.1 | XP_021092649.1 | 3 - 4 | 599 | 72 | ||
More Information
Set Membership: |
|

Larp6
Genomic Map