Assay Details
| Gene Symbol: LOC101707360 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850338.2 | XP_004850395.1 | 14 - 15 | 2227 | 63 | |
| XM_004850339.2 | XP_004850396.1 | 13 - 14 | 2167 | 63 | ||
| XM_021262849.1 | XP_021118508.1 | 14 - 15 | 2306 | 63 | ||
| XM_021262850.1 | XP_021118509.1 | 14 - 15 | 2303 | 63 | ||
| XM_021262851.1 | XP_021118510.1 | 13 - 14 | 2246 | 63 | ||
| XM_021262852.1 | XP_021118511.1 | 13 - 14 | 2243 | 63 | ||
| XM_021262853.1 | XP_021118512.1 | 14 - 15 | 2306 | 63 | ||
| XM_021262854.1 | XP_021118513.1 | 14 - 15 | 2306 | 63 | ||
| XM_021262855.1 | XP_021118514.1 | 14 - 15 | 2306 | 63 | ||
| XM_021262856.1 | XP_021118515.1 | 7 - 8 | 751 | 63 | ||
More Information
Set Membership: |
|

LOC101707360
Genomic Map