Assay Details
| Gene Symbol: Aldh8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004871506.2 | XP_004871563.1 | 6 - 7 | 1900 | 64 | |
| XM_004871507.2 | XP_004871564.1 | 5 - 6 | 1772 | 64 | ||
More Information
Set Membership: |
|

Aldh8a1
Genomic Map