Assay Details
| Gene Symbol: Angpt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869583.3 | XP_004869640.1 | 7 - 8 | 2098 | 60 | |
| XM_013067370.2 | XP_012922824.1 | 7 - 8 | 1105 | 60 | ||
| XM_021248190.1 | XP_021103849.1 | 7 - 8 | 1873 | 60 | ||
More Information
Set Membership: |
|

Angpt2
Genomic Map