Assay Details
| Gene Symbol: Stk17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004868946.2 | XP_004869003.1 | 7 - 8 | 1053 | 71 | |
| XM_021247734.1 | XP_021103393.1 | 7 - 8 | 902 | 71 | ||
More Information
Set Membership: |
|

Stk17b
Genomic Map