Assay Details
| Gene Symbol: Eif2s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867922.2 | XP_004867979.1 | 5 - 6 | 721 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif2s2| Gene Symbol: Eif2s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867922.2 | XP_004867979.1 | 5 - 6 | 721 | 74 | |
Set Membership: |
|