Assay Details
| Gene Symbol: LOC101717854 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846414.3 | XP_004846471.1 | 1 - 2 | 362 | 72 | |
| XM_004846416.3 | XP_004846473.1 | 1 - 2 | 366 | 72 | ||
| XM_004846417.3 | XP_004846474.1 | 1 - 2 | 365 | 72 | ||
More Information
Set Membership: |
|

LOC101717854
Genomic Map